Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHR Antibody, Novus Biologicals™
SDP

Catalog No. NB100128SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100128SS 0.25 mL
NB100128 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100128SS Supplier Novus Biologicals Supplier No. NB100128SS
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 13 publications

AHR Polyclonal specifically detects AHR in Human, Mouse, Rat samples. It is validated for Chemotaxis, ChIP Assay, EMSA, Gel Super Shift Assays, Gel Supershift, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen AHR
Applications Chemotaxis, ChIP Assay, EMSA, Gel Supershift, Gel Supershift, Immunocytochemistry, Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot reported in scientific literature (PMID 31351871; 24302727; 3999131), Chromatin Immunoprecipitation reported in scientific literature (PMID 3999131), Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 - 1:200, Gel Super Shift Assays
Formulation Whole antisera with No Preservative
Gene Alias Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76
Gene Symbols AHR
Host Species Goat
Immunogen N-terminal sequence of Aryl hydrocarbon Receptor purified from C57BL/6J mice. [UniProt# P30561]
Purification Method Unpurified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 196
Test Specificity Human and mouse. Rat reactivity reported in the scientific literature (PMID: 18515748).
Target Species Human, Mouse, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.