Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHR Antibody, Novus Biologicals™
SDP

Goat Polyclonal Antibody has been used in 13 publications

$192.47 - $529.95

Specifications

Antigen AHR
Dilution Western Blot reported in scientific literature (PMID 31351871; 24302727; 3999131), Chromatin Immunoprecipitation reported in scientific literature (PMID 3999131), Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 - 1:200, Gel Super Shift Assays
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100128SS
SDP
View Documents
Novus Biologicals
NB100128SS
0.25 mL
Each for $192.47
Only null left
Add to Cart
 
NB100128
SDP
View Documents
Novus Biologicals
NB100128
1 mL
Each for $529.95
Only null left
Add to Cart
 
Description

Description

AHR Polyclonal specifically detects AHR in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Gel Super Shift Assays.
Specifications

Specifications

AHR
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
Goat
Apoptosis, Breast Cancer, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators
Whole antisera with No Preservative
196
N-terminal sequence of Aryl hydrocarbon Receptor purified from C57BL/6J mice. [UniProt# P30561]
Unpurified
Human and mouse. Rat reactivity reported in the scientific literature (PMID: 18515748).
Western Blot reported in scientific literature (PMID 31351871; 24302727; 3999131), Chromatin Immunoprecipitation reported in scientific literature (PMID 3999131), Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 - 1:200, Gel Super Shift Assays
Polyclonal
Antisera
RUO
Human, Mouse, Rat
Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76
AHR
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.