Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHR Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1002289 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002289 0.1 mL
NB1002289SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1002289 Supplier Novus Biologicals Supplier No. NB1002289
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

AHR Polyclonal specifically detects AHR in Guinea Pig, Human, Mouse, Rat samples. It is validated for Chemotaxis, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Simple Western, Western Blot.

Specifications

Antigen AHR
Applications Chemotaxis, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Simple Western 1:200, Flow Cytometry 2.5 ug/ml, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P35869
Gene Alias Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76
Gene Symbols AHR
Host Species Rabbit
Immunogen Bacterially expressed human Aryl hydrocarbon Receptor (C-terminus). [UniProt# P35869]
Molecular Weight of Antigen 96 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 196
Test Specificity Human, Mouse and Guinea Pig. Rat reactivity reported in scientific literature (PMID: 23887904).
Target Species Guinea Pig, Mouse, Human, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.