Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHR Antibody (RPT9), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB30515A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB30515A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB30515A350 Supplier Novus Biologicals Supplier No. NB300515AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AHR Monoclonal specifically detects AHR in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen AHR
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RPT9
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76
Gene Symbols AHR
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues R(12) K R R K P(17) V(22) K P I P A E G I K(31) of mouse AhR.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Apoptosis, Breast Cancer, Cancer, Chromatin Research, Hypoxia, Innate Immunity, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 196.0
Test Specificity Detects the Aryl Hydrocarbon Receptor (AHR).
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.