Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHSA1 Antibody (OTI1D2), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP271608G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271608G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271608G Supplier Novus Biologicals Supplier No. NBP271608G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AHSA1 Monoclonal specifically detects AHSA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.

Specifications

Antigen AHSA1
Applications Western Blot, Immunohistochemistry, Immunoassay
Classification Monoclonal
Clone OTI1D2
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Knockout Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1
Gene Symbols AHSA1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-203 of human AHSA1 (NP_036243) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10598.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.