Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIBZIP Antibody, Novus Biologicals™
SDP

Catalog No. NB1001015 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001015 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001015 Supplier Novus Biologicals Supplier No. NB1001015
Add to Cart
Add to Cart

Goat Polyclonal Antibody

AIBZIP Polyclonal specifically detects AIBZIP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen AIBZIP
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 2 ug/ml, Immunohistochemistry 2 - 4 ug/ml, Immunohistochemistry-Paraffin 2 - 4 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q8TEY5
Gene Alias AIbZIP, AIBZIPcAMP-responsive element-binding protein 4, Androgen-induced basic leucine zipper protein, ATCE1JAL, Attaching to CRE-like 1, cAMP responsive element binding protein 1, cAMP responsive element binding protein 3-like 4, CREB3, CREB4CREB-4, cyclic AMP-responsive element-binding protein 3-like protein 4, Cyclic AMP-responsive element-binding protein 4, hJALcAMP-responsive element-binding protein 3-like protein 4, Tisp40, Transcript induced in spermiogenesis protein 40
Gene Symbols CREB3L4
Host Species Goat
Immunogen Peptide with sequence C-PSGRIRSVLHADEM corresponding to C-Terminus according to NP_570968.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 148327
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.