Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AICDA Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB215721 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB215721 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB215721 Supplier Novus Biologicals Supplier No. NBP177235AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AICDA Polyclonal antibody specifically detects AICDA in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen AICDA
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias activation-induced cytidine deaminase, AIDCytidine aminohydrolase, ARP2, CDA2, HIGM2EC 3.5.4.5, integrated into Burkitt's lymphoma cell line Ramos
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 14 amino acids near the carboxy terminus of human AID. The immunogen is located within the last 50 amino acids of AID. Amino Acid Squence: EVDDLRDAFRTLGL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Epigenetics, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 57379
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.