Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ AIDA

Click to view available options
Quantity:
10 mg
50 mg
Description
A relatively potent and selective antagonist of group I metabotropic glutamate receptors (mGlu1a), having no effect on group II (mGlu2) or group III (mGlu4) receptors expressed individually in baby hamster kidney cells. Has no effect on ionotropic glutamate receptors. Centrally active following systemic administration in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 221.21 |
| Purity | >98% |
| Molecular Weight (g/mol) | 220.21 |
| Chemical Name or Material | AIDA |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction