Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIF-1/Iba1 Antibody (1022-5), Novus Biologicals™
SDP

Catalog No. NBP150156 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP150156 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP150156 Supplier Novus Biologicals Supplier No. NBP150156

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

AIF-1/Iba1 Monoclonal specifically detects AIF-1/Iba1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoassay.

Specifications

Antigen AIF-1/Iba1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
Classification Monoclonal
Clone 1022-5
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:50, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50, Immunoassay 1:50
Gene Accession No. P55008
Gene Alias AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1
Gene Symbols AIF1
Host Species Mouse
Immunogen This AIF-1/Iba1 Antibody (1022-5) was made using a portion of the human IBA1 gene.
Molecular Weight of Antigen 16.7 kDa
Purification Method Protein A or G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 199
Test Specificity The monoclonal AIF-1/Iba1 Antibody (1022-5) is cross reactive with rat Allograft-inflammatory factor-1 (rat AIF-1). Human and rat amino acids sequences are 98% homologous.
Target Species Human, Rat
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.