Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIF-1/Iba1 Antibody (145), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB038432 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038432 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038432 Supplier Novus Biologicals Supplier No. NBP290272AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

AIF-1/Iba1 Monoclonal specifically detects AIF-1/Iba1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen AIF-1/Iba1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 145
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human AIF-1/Iba1 (P55008-1; Met1-Pro147).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 199
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.