Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIF-1/Iba1 Antibody, Novus Biologicals™
SDP

Catalog No. NB780588 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
20μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB780588 20μg
NB1001028 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB780588 Supplier Novus Biologicals Supplier No. NB100102820UG
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 192 publications

AIF-1/Iba1 Polyclonal specifically detects AIF-1/Iba1 in Human, Mouse, Rat, Canine, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount, Knockout Validated, Dual RNAscope ISH-IHC.

Specifications

Antigen AIF-1/Iba1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ELISA, Immunohistochemistry (Free Floating), Immunoassay
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 1.0 ug/mL, Immunohistochemistry 4 - 6 ug/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 4 - 6 ug/mL, Immunohistochemistry-Frozen, Peptide ELISA Detection Limit 1:128000, Immunohistochemistry Free-Floating, Immunohistochemistry (Whole-Mount), Knockout Validated, Dual RNAscope ISH-IHC
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P55008
Gene Alias AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1
Gene Symbols AIF1
Host Species Goat
Immunogen This AIF-1/Iba1 Antibody was developed against a peptide with sequence C-TGPPAKKAISELP, from the C Terminus of the protein sequence according to NP_116573.1; NP_001614.3.
Molecular Weight of Antigen 16.8 kDa
Purification Method Affinity Purified
Quantity 20μg
Regulatory Status RUO
Research Discipline Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 199
Test Specificity This AIF-1/Iba1 Antibody is expected to recognize isoform 1 (NP_116573.1) and isoform 3 (NP_001614.3). AIF-1/Iba1 is thought to be involved in negative regulation of growth of vascular smooth muscle cells, which contributes to the anti-inflammatory response to vessel wall trauma.
Target Species Human, Mouse, Rat, Canine, Feline
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.