Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIFM3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146149 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146149 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146149 Supplier Novus Biologicals Supplier No. NBP146149
Add to Cart
Add to Cart

Goat Polyclonal Antibody

AIFM3 Polyclonal specifically detects AIFM3 in Bovine, Canine, Human, Mouse, Rat samples. It is validated for Peptide ELISA.

Specifications

Antigen AIFM3
Applications Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_653305.1
Gene Alias AIFLFLJ30473, apoptosis-inducing factor 3, apoptosis-inducing factor like, apoptosis-inducing factor, mitochondrion-associated, 3, Apoptosis-inducing factor-like protein, FLJ45137
Gene Symbols AIFM3
Host Species Goat
Immunogen Peptide with sequence C-TARHFHTEERLSTPH corresponding to internal region according to NP_001018070.1, NP_001139760.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150209
Test Specificity This antibody is expected to recognize all isoforms (NP_653305.1; NP_001018070.1, NP_001139760.1).
Target Species Bovine, Canine, Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.