Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIM2 Antibody (10M2B3), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP227355V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227355V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227355V Supplier Novus Biologicals Supplier No. NBP227355V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AIM2 Monoclonal specifically detects AIM2 in Human samples. It is validated for Western Blot.

Specifications

Antigen AIM2
Applications Western Blot
Classification Monoclonal
Clone 10M2B3
Conjugate DyLight 405
Dilution Western Blot
Gene Accession No. O14862
Gene Alias absent in melanoma 2PYHIN4, interferon-inducible protein AIM2
Gene Symbols AIM2
Host Species Mouse
Immunogen This antibody was raised against recombinant human AIM2 protein
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9447
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.