Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIM2 Antibody (10M5G5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP22734A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP22734A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP22734A94 Supplier Novus Biologicals Supplier No. NBP227354AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AIM2 Monoclonal specifically detects AIM2 in Human samples. It is validated for Western Blot.

Specifications

Antigen AIM2
Applications Western Blot
Classification Monoclonal
Clone 10M5G5
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias absent in melanoma 2PYHIN4, interferon-inducible protein AIM2
Gene Symbols AIM2
Host Species Mouse
Immunogen This antibody was raised against AIM2
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9447.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.