Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aiolos/IKZF3 Antibody (914821), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB295604 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295604 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295604 Supplier Novus Biologicals Supplier No. FAB8625R

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Aiolos/IKZF3 Monoclonal antibody specifically detects Aiolos/IKZF3 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Aiolos/IKZF3
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 914821
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, zinc finger protein Aiolos, ZNFN1A3subfamily 1A, 3 (Aiolos)
Host Species Mouse
Immunogen E. coli-derived recombinant human Aiolos/IKZF3, Met1-Gly54, Accession # Q9UKT9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 22806
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.