Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aiolos/IKZF3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216938 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216938 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216938 Supplier Novus Biologicals Supplier No. NBP216938
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aiolos/IKZF3 Polyclonal specifically detects Aiolos/IKZF3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Aiolos/IKZF3
Applications Western Blot
Classification Polyclonal
Concentration 0.74mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Simple Western reported by internal validation
Formulation 0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9UKT9
Gene Alias AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, zinc finger protein Aiolos, ZNFN1A3subfamily 1A, 3 (Aiolos)
Gene Symbols IKZF3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human IKZF3. The exact sequence is proprietary.
Molecular Weight of Antigen 58 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 22806
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.