Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aiolos/IKZF3 Antibody (OTI7E11), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB271698A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271698A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271698A48 Supplier Novus Biologicals Supplier No. NBP271698AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aiolos/IKZF3 Monoclonal specifically detects Aiolos/IKZF3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Aiolos/IKZF3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7E11
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, zinc finger protein Aiolos, ZNFN1A3subfamily 1A, 3 (Aiolos)
Gene Symbols IKZF3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-301 of human Aiolos/IKZF3 (NP_036613) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 22806.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.