Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIPL1 Antibody (OTI3B4), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP27177A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27177A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27177A94 Supplier Novus Biologicals Supplier No. NBP271775AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AIPL1 Monoclonal specifically detects AIPL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen AIPL1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3B4
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AIPL2, aryl hydrocarbon receptor interacting protein-like 1, aryl hydrocarbon receptor-interacting protein-like 1, aryl-hydrocarbon-interacting protein-like 1, LCA4
Gene Symbols AIPL1
Host Species Mouse
Immunogen Full length recombinant protein of human AIPL1 (NP_055151) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 23746.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.