Learn More
Corning™ PYREX™ Air Reflux Condenser With 14/10 Threaded Joint
For use with Pyrex microchemistry kits
$519.00
Specifications
Material Borosilicate Glass
Type Air Reflux Condenser
For Use With (Equipment) Pyrex microchemistry kits
Quantity 6/Cs.
View More Specs
Products 1
Catalog Number Mfr. No. Standard Taper Joint No Price Quantity & Availability  
Catalog Number Mfr. No. Standard Taper Joint No Price Quantity & Availability  
11-850-107
View Documents
Corning™
285014
14/10 Bottom, 7/10 Top joint
Case of 6 for $519.00
Only null left
Add to Cart
Description
Description
Replacement part for 6949M series of PYREX microchemistry kits.
Specifications
Specifications
Borosilicate Glass
Air Reflux Condenser
Pyrex microchemistry kits
6/Cs.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.