Learn More
AirClean™ Systems Polypropylene Fume Hood Accessory, Base Cabinets
Base cabinets with variety of shelving options are used with AirClean Polypropylene Fume Hoods.
$3729.00 - $4981.00
| Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Width (English) | Height Overall (English) | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Width (English) | Height Overall (English) | Price | Quantity & Availability | ||
36-100-0103
![]() |
AirClean™ Systems
ACA2062 |
Vented | Polypropylene Fume Hood | 72 in. | 32 in. |
Each for $3,729.00
|
|
||
36-100-0124
![]() |
AirClean™ Systems
ACA1083T |
Unvented | AC8000S, ACPT8000S | 96 in. | 36 in. |
Each for $4,981.00
|
|
||
36-100-0098
![]() |
AirClean™ Systems
ACA1042ST |
Vented | AC4000S | 36 in. | N/A | N/A | N/A | ||
Description
- Ideal for storage of corrosive chemicals
- May be vented into the corresponding hood to prevent excessive fume buildup
Specifications
| Polypropylene |