Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIRE Mouse anti-Human, Porcine, Clone: 1A3C3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | AIRE |
---|---|
Clone | 1A3C3 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6695
![]() ![]() |
Proteintech Group Inc
662621IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The autoimmune regulator (AIRE) is a protein that in humans is encoded by the AIRE gene. This gene encodes a transcriptional regulator that forms nuclear bodies and interacts with the transcriptional coactivator CREB binding protein. The encoded protein plays an important role in immunity by regulating the expression of autoantigens and negative selection of autoreactive T-cells in the thymus. Mutations in this gene cause the rare autosomal-recessive systemic autoimmune disease termed autoimmune polyendocrinopathy with candidiasis and ectodermal dystrophy (APECED).Specifications
AIRE | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AIRE, AIRE1, APECED, APECED protein, APS1, APSI, autoimmune regulator, PGA1 | |
AIRE | |
IgG1 | |
Protein G | |
Antibody |
1A3C3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
O43918 | |
100621659, 326 | |
AIRE Fusion Protein Ag19103 | |
Primary | |
-20°C | |
AIRE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title