Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AIRE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586368
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586368 Supplier Invitrogen™ Supplier No. PA586368
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

AIRE is a transcriptional regulator protein that binds to DNA as a dimer and tetramer, but not as a monomer, and may be involved in the regulation of gene expression. A nuclear protein, AIRE is a 545 amino acid protein with an HSR domain, two PHD type zinc fingers, and a SAND domain, and may be present in soluble high-molecular-weight complexes. It may form homodimer, homotetramer and also nuclear bodies and interacts with the transcriptional coactivator CBP and CREBBP. Three isoforms of AIRE are reported and one of them is predicted to be a candidate for nuclear-mediated decay (NMD). AIRE is involved in immune regulation and is known to mediate E3 ubiquitin ligase activity. Defects in AIRE are responsible for the rare autosomal-recessive systemic autoimmune disease termed autoimmune polyendocrinopathy-candidiasis-ectodermal dystrophy (APECED).
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIRE
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Aire
Gene Accession No. O43918, Q9Z0E3
Gene Alias Aire; AIRE protein; AIRE1; APECED; APECED protein; APECED protein homolog; APS1; APSI; Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein; autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein homolog; Autoimmune regulator; autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy); autoimmune regulator AIRE1a; LOW QUALITY PROTEIN: autoimmune regulator; OTTHUMP00000109529; PGA1
Gene Symbols Aire
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids 101-150 of Human AIRE.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11634, 294328, 326
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.