Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ajuba Antibody, Novus Biologicals™
SDP

Catalog No. NB600468 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600468 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600468 Supplier Novus Biologicals Supplier No. NB600468
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ajuba Polyclonal specifically detects Ajuba in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Ajuba
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Concentration 1.67 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2500, ELISA 1:20000-1:80000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q96IF1, Q96IF1, Q96IF1, Q96IF1
Gene Alias Ajuba, jub, ajuba homolog (Xenopus laevis), MGC15563, protein ajuba
Gene Symbols AJUBA
Host Species Rabbit
Immunogen This affinity purified Ajuba Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding aa 224-239 of Human Ajuba. (Uniprot: Q96IF1)
Molecular Weight of Antigen 56.93 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 84962
Test Specificity A BLAST analysis was used to suggest reactivity with this protein from human, rat, dog, mouse and chimpanzee based on 100% identity for the immunogen sequence. Cross reactivity with Ajuba protein homologues from other sources has not been determined.
Reconstitution Reconstitute with 50 ul of distilled water to a final concentration of 1.0 mg/ml.
Target Species Human, Mouse, Rat, Canine, Primate
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.