Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AK3L1 Antibody (010), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB316932 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316932 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316932 Supplier Novus Biologicals Supplier No. NBP290093MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

AK3L1 Monoclonal antibody specifically detects AK3L1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen AK3L1
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 010
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias adenylate kinase 3, Adenylate kinase 3-like, adenylate kinase 3-like 1, adenylate kinase 4, AK3adenylate kinase isoenzyme 4, mitochondrial, AK3L1, AK3L2, ATP-AMP transphosphorylase, EC 2.7.4, EC 2.7.4.3, GTP:AMP phosphotransferase, MGC166959, mitochondrial adenylate kinase-3, nucleoside-triphosphate-adenylate kinase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human AK3L1 (Uniprot#: P27144-1; Ala 2-Tyr 223).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 205
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.