Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AK3L1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB097541 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB097541 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB097541 Supplier Novus Biologicals Supplier No. NBP297592AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AK3L1 Polyclonal specifically detects AK3L1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen AK3L1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias adenylate kinase 3, Adenylate kinase 3-like, adenylate kinase 3-like 1, adenylate kinase 4, AK3adenylate kinase isoenzyme 4, mitochondrial, AK3L1, AK3L2, ATP-AMP transphosphorylase, EC 2.7.4, EC 2.7.4.3, GTP:AMP phosphotransferase, MGC166959, mitochondrial adenylate kinase-3, nucleoside-triphosphate-adenylate kinase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human AK3L1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 205
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.