Learn More
Invitrogen™ AK6 Monoclonal Antibody (B1-F4)

Description
AK6 Monoclonal Antibody for ICC/IF, Western Blot
Specifications
Specifications
| Antigen | AK6 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | B1-F4 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% Glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | AK6 |
| Gene Accession No. | Q5EB68, Q8VCP8, Q9Y3D8 |
| Gene Alias | 2810046E22Rik; 4921516M08Rik; AD-004; adenylate kinase 6; adenylate kinase isoenzyme 6; adenylate kinase isoenzyme 6 {ECO:0000255; Adrenal gland protein AD-004; AK/ATPase; AK/ATPase {ECO:0000255; AK6; AK6 {ECO:0000255; ATP-AMP transphosphorylase 6; CGI-137; CINAP; CIP; coilin interacting nuclear ATPase protein; coilin interacting protein; coilin-interacting nuclear ATPase protein; coilin-interacting nuclear ATPase protein {ECO:0000255; Dual activity adenylate kinase/ATPase; dual activity adenylate kinase/ATPase {ECO:0000255; HAMAP-Rule:MF_03173}; hCINAP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.