Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAIN1 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB163556 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163556 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163556 Supplier Novus Biologicals Supplier No. NBP281698FR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AKAIN1 Polyclonal antibody specifically detects AKAIN1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen AKAIN1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias C18orf42, C18orf42 chromosome 18 open reading frame 42
Host Species Rabbit
Immunogen AKAIN1 antibody was raised against a 16 amino acid peptide from near the center of human AKAIN1. The immunogen is located within the last 50 amino acids of AKAIN1. Amino Acid Squence: VSQESQRREERISDNR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 642597
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.