Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP3 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001411 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001411 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001411 Supplier Novus Biologicals Supplier No. NB1001411
Add to Cart
Add to Cart

Goat Polyclonal Antibody

AKAP3 Polyclonal specifically detects AKAP3 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen AKAP3
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. O75969
Gene Alias A kinase (PRKA) anchor protein 3, AKAP110AKAP 110, AKAP-3, A-kinase anchor protein 110 kDa, Cancer/testis antigen 82, CT82A-kinase anchor protein 3, Fibrous sheath protein of 95 kDa, fibrousheathin 1, Fibrousheathin I, Fibrousheathin-1, FSP95A-kinase anchor protein, 110kDa, PRKA3, protein kinase A anchoring protein 3, Protein kinase A-anchoring protein 3, SOB1fibrous sheath protein, 95kDa, Sperm oocyte-binding protein, sperm oocyte-binding protein 1
Gene Symbols AKAP3
Host Species Goat
Immunogen Peptide with sequence C-TPLQLLDWLMVNL corresponding to C-Terminus according to NP_006413.2.
Molecular Weight of Antigen 95 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10566
Test Specificity Expected from sequence similarity: Mouse, Rat, Dog, Pig, Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.