Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP189173 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP189173 0.1 mL
NB430869 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP189173 Supplier Novus Biologicals Supplier No. NBP189173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AKAP3 Polyclonal specifically detects AKAP3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AKAP3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias A kinase (PRKA) anchor protein 3, AKAP110AKAP 110, AKAP-3, A-kinase anchor protein 110 kDa, Cancer/testis antigen 82, CT82A-kinase anchor protein 3, Fibrous sheath protein of 95 kDa, fibrousheathin 1, Fibrousheathin I, Fibrousheathin-1, FSP95A-kinase anchor protein, 110kDa, PRKA3, protein kinase A anchoring protein 3, Protein kinase A-anchoring protein 3, SOB1fibrous sheath protein, 95kDa, Sperm oocyte-binding protein, sperm oocyte-binding protein 1
Gene Symbols AKAP3
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:AQGGRRDARSFVEAAGTTNFPANEPPVAPDESCLKSAPIVGDQEQAEKKDLRSVFFNFIRNLLSETIFKRDQSPEPKVPEQPVKEDRKLCERP
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10566
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.