Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP7 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB043163 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB043163 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB043163 Supplier Novus Biologicals Supplier No. NBP297418DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AKAP7 Polyclonal specifically detects AKAP7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AKAP7
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias A kinase (PRKA) anchor protein 7, AKAP 18, AKAP15, AKAP18A-kinase anchor protein 7 isoform alpha, AKAP-7 isoform gamma, AKAP-7 isoforms alpha and beta, A-kinase anchor protein 18 kDa, A-kinase anchor protein 7, A-kinase anchor protein 7 isoform gamma, A-kinase anchor protein 7 isoforms alpha and beta, A-kinase anchor protein 9 kDa, A-kinase anchor protein, 18-kD, A-kinase anchoring protein 18, protein kinase A anchoring protein 7, Protein kinase A-anchoring protein 7 isoform gamma, Protein kinase A-anchoring protein 7 isoforms alpha/beta
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human AKAP7 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9465
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.