Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP9/CG-NAP Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

$151.31

Specifications

Antigen AKAP9/CG-NAP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-156-0939
Encompass_Preferred
View Documents
Bethyl Laboratories, Inc
A301662AT
20 μL
Each of 1 for $151.31
Only null left
Add to Cart
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

The A-kinase anchor proteins are a group of structurally diverse proteins which have the common function of binding to the regulatory subunit of protein kinase A and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternate splicing of this gene results in at least two isoforms that localize to the centrosome and the Golgi apparatus, and interact with numerous signaling proteins from multiple signal transduction pathways. These signaling proteins include type II protein kinase A, serine/threonine kinase protein kinase N, protein phosphatase 1, protein phosphatase 2a, protein kinase C-epsilon and phosphodiesterase 4D3.
Specifications

Specifications

AKAP9/CG-NAP
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
AKAP9, CG-NAP, PRKA9, YOTIAO, AKAP350, HYPERION, KIAA0803, A kinase (PRKA) anchor protein (yotiao) 9, AKAP120-like protein, kinase N-associated protein, A-kinase anchoring protein 450, A-kinase anchor protein, 350kDa, protein kinase A anchoring protein 9, centrosome- and golgi-localized protein, AKAP450, MU-RMS-40.16A, A-kinase anchor protein 9, AKAP9-BRAF fusion protein
AKAP9
IgG
Antigen affinity chromatography
Antibody
Western Blot
Unconjugated
Rabbit
Human
Q99996
10142
Between 1075 and 1125
Primary
4° C
AKAP9
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.