Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKIRIN1 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB163596 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163596 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163596 Supplier Novus Biologicals Supplier No. NBP281700DL594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AKIRIN1 Polyclonal antibody specifically detects AKIRIN1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen AKIRIN1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias AKIRIN1 akirin 1, C1orf108, RP11-781D11.2, STRF2
Host Species Rabbit
Immunogen AKIRIN1 antibody was raised against a 14 amino acid synthetic peptide near the center of the human AKIRIN1. The immunogen is located within amino acids 40 - 90 of AKIRIN1. Amino Acid Squence: PAPPGSERRLPTPE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 79647
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.