Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Akirin2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB358371 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358371 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358371 Supplier Novus Biologicals Supplier No. NBP176347MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Akirin2 Polyclonal antibody specifically detects Akirin2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen Akirin2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias akirin 2, akirin-2, C6orf166, chromosome 6 open reading frame 166, dJ486L4.2, FBI1, FLJ10342, fourteen-three-three beta interactant 1
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the center of the human Akirin2. The immunogen is located within amino acids 80 - 130 of Akirin2. Amino Acid Squence: ETSFQQTDPCCTSDA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 55122
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.