Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKR1A1 Antibody (OTI6E3), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP271591UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271591UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271591UV Supplier Novus Biologicals Supplier No. NBP271591UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AKR1A1 Monoclonal specifically detects AKR1A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen AKR1A1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI6E3
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias alcohol dehydrogenase, alcohol dehydrogenase [NADP+], Aldehyde reductase, Aldo-keto reductase family 1 member A1, aldo-keto reductase family 1, member A1 (aldehyde reductase), ALDR1, ALRMGC1380, ARM, DD3, dihydrodiol dehydrogenase 3, EC 1.1.1, EC 1.1.1.2, MGC12529
Gene Symbols AKR1A1
Host Species Mouse
Immunogen Full length recombinant protein of human AKR1A1 (NP_006057) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10327.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.