Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKR1B1 Antibody (AKR1B1/7010R), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB157894 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB157894 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB157894 Supplier Novus Biologicals Supplier No. NBP314284JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

AKR1B1 Monoclonal antibody specifically detects AKR1B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen AKR1B1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AKR1B1/7010R
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ADR, Aldehyde reductase, Aldo-keto reductase family 1 member B1, aldo-keto reductase family 1, member B1 (aldose reductase), ALDR1aldose reductase, ALR2, ARaldehyde reductase 1, EC 1.1.1, EC 1.1.1.21, Lii5-2 CTCL tumor antigen, low Km aldose reductase, MGC1804
Host Species Rabbit
Immunogen Recombinant human full-length AKR1B1 protein (Uniprot: P15121)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 231
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.