Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKR1B1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB045243 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045243 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045243 Supplier Novus Biologicals Supplier No. NBP297585B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AKR1B1 Polyclonal specifically detects AKR1B1 in Human samples. It is validated for Western Blot.

Specifications

Antigen AKR1B1
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Formulation PBS
Gene Alias ADR, Aldehyde reductase, Aldo-keto reductase family 1 member B1, aldo-keto reductase family 1, member B1 (aldose reductase), ALDR1aldose reductase, ALR2, ARaldehyde reductase 1, EC 1.1.1, EC 1.1.1.21, Lii5-2 CTCL tumor antigen, low Km aldose reductase, MGC1804
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human AKR1B1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 231
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.