Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKR1C2 Antibody (CPTC-AKR1C2-1), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB012341 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB012341 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB012341 Supplier Novus Biologicals Supplier No. NBP279909AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AKR1C2 Monoclonal specifically detects AKR1C2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AKR1C2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CPTC-AKR1C2-1
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias aldo-keto reductase family 1 member C2, aldo-keto reductase family 1, member C2 (dihydrodiol dehydrogenase 2; bile acidbinding protein; 3-alpha hydroxysteroid dehydrogenase, type III), BABP, Chlordecone reductase homolog HAKRD, DD, DD-2, DD2DD/BABP, DDH2FLJ53800, Dihydrodiol dehydrogenase 2, Dihydrodiol dehydrogenase/bile acid-binding protein, EC 1.1.1,3-alpha-HSD3, EC 1.1.1.213, EC 1.3.1.20, HAKRDAKR1C-pseudo, HBAB, MCDR2, pseudo-chlordecone reductase, Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase, type II dihydrodiol dehydrogenase, Type III 3-alpha-hydroxysteroid dehydrogenase
Gene Symbols AKR1C2
Host Species Mouse
Immunogen Recombinant human full-length AKR1C2 protein (Uniprot: P52895)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.