Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKR7A3 Antibody (2E11), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP197944A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP197944A70 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP197944A70 Supplier Novus Biologicals Supplier No. NBP197944AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AKR7A3 Monoclonal specifically detects AKR7A3 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen AKR7A3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 200000000000
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AFAR2, AFB1 aldehyde reductase 2, AFB1-AR 2, aflatoxin B1 aldehyde reductase 2, aflatoxin B1 aldehyde reductase member 3, aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase)
Gene Symbols AKR7A3
Host Species Mouse
Immunogen Recombinant human AKR7A3 (1-331aa) purified from E. coli
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 22977.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.