Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKR7A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | AKR7A3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
AKR7A3 Polyclonal specifically detects AKR7A3 in Human samples. It is validated for Western Blot.Specifications
AKR7A3 | |
Polyclonal | |
Rabbit | |
O95154 | |
22977 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
AFAR2, AFB1 aldehyde reductase 2, AFB1-AR 2, aflatoxin B1 aldehyde reductase 2, aflatoxin B1 aldehyde reductase member 3, aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase) | |
AKR7A3 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title