Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT Antibody (0002) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB306084 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306084 100 μL
NB306083 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB306084 Supplier Novus Biologicals Supplier No. NBP323519100UL

Rabbit Monoclonal Antibody

AKT Monoclonal antibody specifically detects AKT in Human samples. It is validated for Western Blot

Specifications

Antigen AKT
Applications Western Blot
Classification Monoclonal
Clone 0002
Conjugate Unconjugated
Dilution Western Blot 1:200-1:1000
Formulation 0.2 um filtered solution in PBS
Gene Alias RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the residues around Ser308 of Human phospho-AKT.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cellular Signaling, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 207
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.