Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT, p Ser473 Antibody (545007), FITC, Novus Biologicals™
SDP

Catalog No. NB302531 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302531 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302531 Supplier Novus Biologicals Supplier No. FAB887F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AKT Monoclonal specifically detects AKT in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen AKT
Applications Western Blot
Classification Monoclonal
Clone 545007
Conjugate FITC
Dilution Western Blot, Simple Western
Formulation PBS
Gene Alias RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC
Host Species Mouse
Immunogen Phosphopeptide containing the human Akt (S473) site
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 207
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.