Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP266427 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266427 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266427 Supplier Novus Biologicals Supplier No. NBP266427

Goat Polyclonal Antibody

AKT1 Polyclonal specifically detects AKT1 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen AKT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 - 3 ug/mL, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas
Gene Symbols AKT1
Host Species Goat
Immunogen This AKT1 antibody is made to a peptide with sequence C-EVSLAKPKHR, from the internal region of the protein sequence according to NP_005154.2; NP_001014432.1; NP_001014431.1.
Molecular Weight of Antigen 55.7 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 207
Test Specificity Variants one, two and three encode the same protein (NP_005154, NP_001014432, NP_001014431). No cross-reactivity expected with AKT2 or AKT3.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.