Learn More
Description
Specifications
Specifications
| Antigen | AKT1 (p Ser473) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 17F6.B11 |
| Concentration | LYOPH |
| Conjugate | Biotin |
| Dilution | Western Blot 1:500-1:3000, Flow Cytometry 1:10-1:1000, ELISA 1:20000, Immunohistochemistry 20 ug/ml, Immunocytochemistry/Immunofluorescence 1:500-1:3000, Immunohistochemistry-Paraffin 20 ug/ml |
| Gene Accession No. | P31749 |
| Gene Alias | AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas |
| Gene Symbols | AKT1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
