Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1, p Thr308 Antibody (18F3.H11), Novus Biologicals™
SDP

Catalog No. NBP169924 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP169924 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP169924 Supplier Novus Biologicals Supplier No. NBP169924
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AKT1 (p Thr308) Monoclonal antibody specifically detects AKT1 (p Thr308) in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin).

Specifications

Antigen AKT1 (p Thr308)
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 18F3.H11
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Flow Cytometry 1:1:10 - 1:1000, ELISA 1:20000, Immunohistochemistry 20 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 20 ug/ml
Gene Accession No. P31749
Gene Alias AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas
Gene Symbols AKT1
Host Species Mouse
Immunogen AKT1 [p Thr308] Antibody (18F3.H11) was produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding T308 of human AKT11 protein. (Uniprot: P31749)
Molecular Weight of Antigen 55.7 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 207
Test Specificity This antibody is specific for human and mouse AKT protein phosphorylated at T308. A BLAST analysis was used to suggest cross-reactivity with AKT pT308 from most vertebrate species sources based on 100% homology with the immunizing sequence. Cross-reactivity with AKT from other sources has not been determined. Cross-reactivity with AKT2 and AKT3 will likely occur.
Target Species Human, Mouse, Rat, Monkey, Vertebrates
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.