Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT2 Antibody (95C657.1.2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056676
Description
AKT2 Monoclonal specifically detects AKT2 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
| AKT2 | |
| Monoclonal | |
| Unconjugated | |
| P31751 | |
| AKT2 | |
| This antibody was generated by immunizing mice with synthetic peptide VANSLKQRAPGEDPMDYK (amino acids 106-123) of human AKT2. | |
| 0.2 mL | |
| Apoptosis, Cancer, Hypoxia, mTOR Pathway, Protein Kinase, Signal Transduction | |
| 208 | |
| Human | |
| Ascites |
| Western Blot | |
| 95C657.1.2 | |
| Western Blot 1:500-1:1000 | |
| EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| AKT2 (95C657.1.2) | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction