Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT2 Antibody (95C657.1.2), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $426.00

Specifications

Antigen AKT2
Clone 95C657.1.2
Dilution Western Blot 1:500-1:1000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056676T
SDP
View Documents
Novus Biologicals
NB10056676SS
0.05 mL
Each for $206.50
Only null left
Add to Cart
 
NB10056676
SDP
View Documents
Novus Biologicals
NB10056676
0.2 mL
Each for $426.00
Only null left
Add to Cart
 
Description

Description

AKT2 Monoclonal specifically detects AKT2 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

AKT2
Western Blot 1:500-1:1000
Monoclonal
Ascites
RUO
Human
EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2
AKT2
IgM
Unpurified
AKT2 (95C657.1.2)
95C657.1.2
Western Blot
Unconjugated
Mouse
Apoptosis, Cancer, Hypoxia, mTOR Pathway, Protein Kinase, Signal Transduction
P31751
208
This antibody was generated by immunizing mice with synthetic peptide VANSLKQRAPGEDPMDYK (amino acids 106-123) of human AKT2.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.