Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT3 Antibody (199822), HRP, Novus Biologicals™
SDP

Catalog No. NB349900 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349900 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349900 Supplier Novus Biologicals Supplier No. FAB14631H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AKT3 Monoclonal antibody specifically detects AKT3 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen AKT3
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 199822
Conjugate HRP
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias Akt-3, EC 2.7.11, EC 2.7.11.1, PKB gamma, PKBGDKFZp434N0250, PRKBG, v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma)
Host Species Mouse
Immunogen E. coli-derived recombinant human Akt3, Arg23-Ala101, Accession # NP_005456
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10000
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.