Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT3 Antibody (66C1247), Novus Biologicals™
SDP

Catalog No. NB6001298SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001298SS 0.025 mg
NB6001298 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB6001298SS Supplier Novus Biologicals Supplier No. NB6001298SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AKT3 Monoclonal specifically detects AKT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen AKT3
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 66C1247
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence
Gene Accession No. Q9Y243
Gene Alias Akt-3, EC 2.7.11, EC 2.7.11.1, PKB gamma, PKBGDKFZp434N0250, PRKBG, v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma)
Gene Symbols AKT3
Host Species Mouse
Immunogen This monoclonal antibody was raised against a synthetic peptide corresponding to amino acid residues 119-136 (CSPTSQIDNIGEEEMDAS) of human Akt3, GenBank no. gi 4574744 gb AAD24196.1 AF135794_1. This sequence is identical in human, mouse, rat, cow, dog, and chicken.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10000
Test Specificity Cross-reacts with Human, Mouse and Rat. Not yet tested in other species.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.