Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aflatoxin B1 Antibody (6A10) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB600443 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600443 0.1 mL
NB600443SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600443 Supplier Novus Biologicals Supplier No. NB600443
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 21 publication

Aflatoxin B1 Monoclonal specifically detects Aflatoxin B1 in Human, All Species samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.

Specifications

Antigen Aflatoxin B1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6A10
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 5 ug/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100. Use reported in scientific literature (PMID 24114584), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24391771), Immunohistochemistry-Frozen, Dot Blot reported in scientific literature (PMID 19524575)
Gene Alias AFB1, AFB1-AR1, Aldoketoreductase 7
Host Species Mouse
Immunogen Aflatoxin B1 DNA (against the midazole ring-opened persistent form of the major N-7 guanine adduct of AFB1).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Test Specificity All species. There is no cross-reactivity with other carcinogen-DNA adducts.
Target Species Human, All species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.