Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aflatoxin B1 Antibody (6A10) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 21 publication

$206.50 - $499.50

Specifications

Antigen Aflatoxin B1
Clone 6A10
Concentration 1.0 mg/ml
Dilution Western Blot 1:500-1:2000, ELISA 5 ug/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100. Use reported in scientific literature (PMID 24114584), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24391771), Immunohistochemistry-Frozen, Dot Blot reported in scientific literature (PMID 19524575)
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600443SS
SDP
View Documents
Novus Biologicals
NB600443SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB600443
SDP
View Documents
Novus Biologicals
NB600443
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Aflatoxin B1 Monoclonal specifically detects Aflatoxin B1 in Human, All Species samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.
Specifications

Specifications

Aflatoxin B1
1.0 mg/ml
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer
AFB1, AFB1-AR1, Aldoketoreductase 7
IgG1 κ
Protein G purified
All species. There is no cross-reactivity with other carcinogen-DNA adducts.
6A10
Western Blot 1:500-1:2000, ELISA 5 ug/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100. Use reported in scientific literature (PMID 24114584), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24391771), Immunohistochemistry-Frozen, Dot Blot reported in scientific literature (PMID 19524575)
Monoclonal
Purified
RUO
Human, All species
Aflatoxin B1 DNA (against the midazole ring-opened persistent form of the major N-7 guanine adduct of AFB1).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.